Thermo Scientific Chemicals

2-Naphthoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14046.30
250 g, Each
Thermo Scientific Chemicals

2-Naphthoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14046.30
250 g, Each
Quantity
Catalog number: A14046.30
also known as A14046-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2243-83-6
IUPAC Name
naphthalene-2-carbonyl chloride
Molecular Formula
C11H7ClO
InChI Key
XNLBCXGRQWUJLU-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C2C=CC=CC2=C1
Appearance (Color)
White to cream to yellow to brown to pale grey
Form
Fused solid or crystalline chunks or lumpy solid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text