Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.
Solubility
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene, chloroform, and hydronaphthalenes. Insoluble in water.
Notes
Store in room temperature. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
RUO – Research Use Only
H.A. Azab; S.A. El-Korashy; Z.M. Anwar; B.H.M. Hussein; G.M. Khairy. Synthesis and fluorescence properties of Eu-anthracene-9-carboxylic acid towards N-acetyl amino acids and nucleotides in different solvents. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2010, 75(1), 21-27.
Michael J. Welsh. Anthracene-9-carboxylic acid inhibits an apical membrane, chloride conductance in canine tracheal epithelium. The Journal of Membrane Biology. 1984, 78(1), 61-71.