Thermo Scientific Chemicals

Anthracene-9-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14049.18
50 g, Each
Thermo Scientific Chemicals

Anthracene-9-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14049.18
50 g, Each
Quantity
Catalog number: A14049.18
also known as A14049-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
723-62-6
IUPAC Name
anthracene-9-carboxylic acid
Molecular Formula
C15H10O2
InChI Key
XGWFJBFNAQHLEF-UHFFFAOYSA-N
SMILES
OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
Appearance (Color)
Yellow to yellow-green
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Clarity
0.5% w/v in ethanol: clear solution

Description

Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

Solubility
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene, chloroform, and hydronaphthalenes. Insoluble in water.

Notes
Store in room temperature. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text