Thermo Scientific Chemicals

1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals

Catalog number: A14050.0E
2500 g, Each
Thermo Scientific Chemicals

1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals

Catalog number: A14050.0E
2500 g, Each
Quantity
Catalog number: A14050.0E
also known as A14050-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
637-59-2
IUPAC Name
(3-bromopropyl)benzene
Molecular Formula
C9H11Br
InChI Key
XMZQWZJMTBCUFT-UHFFFAOYSA-N
SMILES
BrCCCC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5440-1.5480 @ 20?C

Description

1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

Solubility
Soluble in water (partly). Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text