Thermo Scientific Chemicals

Quinoxaline, 98+%, Thermo Scientific Chemicals

Catalog number: A14055.36
500 g, Each
Thermo Scientific Chemicals

Quinoxaline, 98+%, Thermo Scientific Chemicals

Catalog number: A14055.36
500 g, Each
Quantity
Catalog number: A14055.36
also known as A14055-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-19-0
IUPAC Name
quinoxaline
Molecular Formula
C8H6N2
InChI Key
XSCHRSMBECNVNS-UHFFFAOYSA-N
SMILES
C1=CC=C2N=CC=NC2=C1
Appearance (Color)
Orange to red to dark brown to brown
Form
Crystals or powder or crystalline powder or fused solid or needles or liquid as melt
Assay (GC)
≥98.0%

Description

Used as dyes, pharmaceuticals and antibiotics intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text