Thermo Scientific Chemicals

Benzo[b]thiophene 1,1-dioxide, 98%, Thermo Scientific Chemicals

Catalog number: A14063.14
25 g, Each
Thermo Scientific Chemicals

Benzo[b]thiophene 1,1-dioxide, 98%, Thermo Scientific Chemicals

Catalog number: A14063.14
25 g, Each
Quantity
Catalog number: A14063.14
also known as A14063-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
825-44-5
IUPAC Name
1λ⁶-benzothiophene-1,1-dione
Molecular Formula
C8H6O2S
InChI Key
FRJNKYGTHPUSJR-UHFFFAOYSA-N
SMILES
O=S1(=O)C=CC2=CC=CC=C12
Appearance (Color)
White or cream to pale orange/brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
136-143?C
Form
Powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text