Thermo Scientific Chemicals

4-(4-Hydroxyphenyl)-2-butanone, 98%, Thermo Scientific Chemicals

Catalog number: A14064.22
100 g, Each
Thermo Scientific Chemicals

4-(4-Hydroxyphenyl)-2-butanone, 98%, Thermo Scientific Chemicals

Catalog number: A14064.22
100 g, Each
Quantity
Catalog number: A14064.22
also known as A14064-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5471-51-2
IUPAC Name
4-(4-hydroxyphenyl)butan-2-one
Molecular Formula
C10H12O2
InChI Key
NJGBTKGETPDVIK-UHFFFAOYSA-N
SMILES
CC(=O)CCC1=CC=C(O)C=C1
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or needles
Melting Point (clear melt)
80.0-87.0°C
Appearance (Color)
White to cream

Description

4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text