Thermo Scientific Chemicals

Sodium bismuth oxide, 80%, Thermo Scientific Chemicals

Catalog number: A14073.14
25 g, Each
Thermo Scientific Chemicals

Sodium bismuth oxide, 80%, Thermo Scientific Chemicals

Catalog number: A14073.14
25 g, Each
Quantity
Catalog number: A14073.14
also known as A14073-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12232-99-4
IUPAC Name
sodium bismuthanoylolate
Molecular Formula
BiNaO3
InChI Key
PNYYBUOBTVHFDN-UHFFFAOYSA-N
SMILES
[Na+].[O-][Bi](=O)=O
Appearance (Color)
Yellow to brown
Form
Crystals or powder or crystalline powder
Assay (Redox Titration)
≥78.0%

Description

Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.

Solubility
Soluble in aqueous acid. Insoluble in water.

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text