Thermo Scientific Chemicals

3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture, Thermo Scientific Chemicals

Catalog number: A14075.22
100 g, Each
Thermo Scientific Chemicals

3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture, Thermo Scientific Chemicals

Catalog number: A14075.22
100 g, Each
Quantity
Catalog number: A14075.22
also known as A14075-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
535-87-5
IUPAC Name
5-carboxybenzene-1,3-bis(aminium) dichloride
Molecular Formula
C7H10Cl2N2O2
InChI Key
IGPLRBRBTUNCRT-UHFFFAOYSA-N
SMILES
[Cl-].[Cl-].[NH3+]C1=CC(=CC([NH3+])=C1)C(O)=O
Appearance (Color)
Grey or brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0% to ≤104.0% (dry wt. basis)
Assay (Silylated GC)
≥96.0%
Water Content (Karl Fischer Titration)
≤3.5%

Description

3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text