3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
A.L.Ahmad; B.S.Ooi; J.P.Choudhury. Preparation and characterization of co-polyamide thin film composite membrane from piperazine and 3,5-diaminobenzoic acid. Desalination. 2003, 158,(1-3), 101-108.
Hans R.Kricheldorf; Gerd Lohden. New polymer syntheses, 79. Hyperbranched poly(ester-amide)s based on 3-hydroxybenzoic acid and 3,5-diaminobenzoic acid. Macromolecular Chemistry and Physics. 1995, 196,(6), 1839-1854.