4-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
4-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number A14081.03
also known as A14081-03
Price (USD)/ Each
59.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.50 
Save 9.85 (14%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
59.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.50 
Save 9.85 (14%)
Add to cart
4-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberA14081.03
Price (USD)/ Each
59.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.50 
Save 9.85 (14%)
-
Add to cart
Chemical Identifiers
CAS49584-26-1
IUPAC Name4-cyanobenzene-1-sulfonyl chloride
Molecular FormulaC7H4ClNO2S
InChI KeyDBMFYTQPPBBKHI-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Melting Point (clear melt)105-111?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only