Thermo Scientific Chemicals

Glycine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A14083.0E
2500 g, Each
Thermo Scientific Chemicals

Glycine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A14083.0E
2500 g, Each
Quantity
Catalog number: A14083.0E
also known as A14083-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5680-79-5
IUPAC Name
hydrogen methyl 2-aminoacetate chloride
Molecular Formula
C3H8ClNO2
InChI Key
COQRGFWWJBEXRC-UHFFFAOYSA-N
SMILES
[H+].[Cl-].COC(=O)CN
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

Solubility
Soluble in water (2% w/v) and methanol (50 mg/ml).

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text