Thermo Scientific Chemicals

Geranyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: A14093.22
100 g, Each
Thermo Scientific Chemicals

Geranyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: A14093.22
100 g, Each
Quantity
Catalog number: A14093.22
also known as A14093-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6138-90-5
IUPAC Name
(6E)-8-bromo-2,6-dimethylocta-2,6-diene
Molecular Formula
C10H17Br
InChI Key
JSCUZAYKVZXKQE-JXMROGBWSA-N
SMILES
CC(C)=CCCC(C)=CCBr
Appearance (Color)
Clear, colourless to yellow
Form
liquid
Assay (Titration ex Bromide)
>95.0%
Proton NMR
Conforms to structure
Refractive Index
1.5010-1.5070

Description

Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text