Thermo Scientific Chemicals

1,4-Dimethoxynaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: A14095.14
25 g, Each
Thermo Scientific Chemicals

1,4-Dimethoxynaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: A14095.14
25 g, Each
Quantity
Catalog number: A14095.14
also known as A14095-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10075-62-4
IUPAC Name
1,4-dimethoxynaphthalene
Molecular Formula
C12H12O2
InChI Key
FWWRTYBQQDXLDD-UHFFFAOYSA-N
SMILES
COC1=C2C=CC=CC2=C(OC)C=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥95.0%
Melting Point (clear melt)
82.0-92.0?C

Description

1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text