Thermo Scientific Chemicals

2-(Chloromethyl)quinoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14102.06
5 g, Each
Thermo Scientific Chemicals

2-(Chloromethyl)quinoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14102.06
5 g, Each
Quantity
Catalog number: A14102.06
also known as A14102-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3747-74-8
IUPAC Name
hydrogen 2-(chloromethyl)quinoline chloride
Molecular Formula
C10H9Cl2N
InChI Key
WDETYCRYUBGKCE-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCC1=CC=C2C=CC=CC2=N1
Form
Powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Appearance (Color)
Cream to pale pink

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text