Thermo Scientific Chemicals

Piperonylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14105.14
25 g, Each
Thermo Scientific Chemicals

Piperonylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14105.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14105.14
also known as A14105-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-53-1
IUPAC Name
2H-1,3-benzodioxole-5-carboxylic acid
Molecular Formula
C8H6O4
InChI Key
VDVJGIYXDVPQLP-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C2OCOC2=C1
Assay (Silylated GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to yellow or cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.0% to ≤102.0%

Description

Piperonylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text