2-(Trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
2-(Trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
25 g
Catalog number A14108.22
also known as A14108-22
Price (USD)/ Each
742.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
742.00
Add to cart
2-(Trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA14108.22
Price (USD)/ Each
742.00
-
Add to cart
Chemical Identifiers
CAS776-04-5
IUPAC Name2-(trifluoromethyl)benzene-1-sulfonyl chloride
Molecular FormulaC7H4ClF3O2S
InChI KeyZIZGWNOAHUCACM-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC=C1S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5010-1.5050 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only