4,4'-Dimethylbenzophenone, 98+%, Thermo Scientific Chemicals
4,4'-Dimethylbenzophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Dimethylbenzophenone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A14111.14
also known as A14111-14
Price (USD)/ Each
151.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
151.00
Add to cart
4,4'-Dimethylbenzophenone, 98+%, Thermo Scientific Chemicals
Catalog numberA14111.14
Price (USD)/ Each
151.00
-
Add to cart
Chemical Identifiers
CAS611-97-2
IUPAC Namebis(4-methylphenyl)methanone
Molecular FormulaC15H14O
InChI KeyZWPWLKXZYNXATK-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)91.0-97.0?C
4,4'-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
Okba Saied; Michel Simard and James D. Wuest. Complexes of Strong Bidentate Lewis Acids Derived from 2,7-Bis(1,1-dimethylethyl)fluorene-1,8-diol. Inorg. Chem. 1998, 37 (11), 2620-2625.