Thermo Scientific Chemicals

2-Bromophenol, 98%, Thermo Scientific Chemicals

Catalog number: A14112.36
500 g, Each
Thermo Scientific Chemicals

2-Bromophenol, 98%, Thermo Scientific Chemicals

Catalog number: A14112.36
500 g, Each
Quantity
Catalog number: A14112.36
also known as A14112-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-56-7
IUPAC Name
2-bromophenol
Molecular Formula
C6H5BrO
InChI Key
VADKRMSMGWJZCF-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1Br
Appearance (Color)
Clear colorless to yellow or pale orange
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5870-1.5910 @ 20?C
Form
Liquid

Description

2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

Solubility
Soluble in chloroform and ether. Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text