Thermo Scientific Chemicals

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific Chemicals

Catalog number: A14121.18
50 g, Each
Thermo Scientific Chemicals

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific Chemicals

Catalog number: A14121.18
50 g, Each
Quantity
Catalog number: A14121.18
also known as A14121-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13209-15-9
IUPAC Name
1,2-bis(dibromomethyl)benzene
Molecular Formula
C8H6Br4
InChI Key
LNAOKZKISWEZNY-UHFFFAOYSA-N
SMILES
BrC(Br)C1=CC=CC=C1C(Br)Br
Appearance (Color)
White to cream to pale brown to pale gray
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
111.0-120.0?C

Description

α,α,α',α'-Tetrabromo-o-xylene reacts with 5endo,6endo-bis-chlormethyl-norborn-2-en to obtain trans/cis-2,3-bis(chloromethyl)-1,4-methano-1,2,3,4-tetrahydroanthracene. Further, it is involved in the preparation of 1-(3-butenyl)-2-(deuteriomethyl)benzene by reacting with allylmagnesium bromide. In addition this, it is used to prepare exo-benzocyclobuteno-1,2,3,4-tetrahydro-1,4-methanoanthracene by reacting with exo-1,4,4a,8b-tetrahydro-1,4-methanobiphenylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α,α,α′,α′-Tetrabromo-o-xylene reacts with 5endo,6endo-bis-chlormethyl-norborn-2-en to obtain trans/cis-2,3-bis(chloromethyl)-1,4-methano-1,2,3,4-tetrahydroanthracene. Further, it is involved in the preparation of 1-(3-butenyl)-2-(deuteriomethyl)benzene by reacting with allylmagnesium bromide. In addition this, it is used to prepare exo-benzocyclobuteno-1,2,3,4-tetrahydro-1,4-methanoanthracene by reacting with exo-1,4,4a,8b-tetrahydro-1,4-methanobiphenylene.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text