Geltrex™ LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix
Geltrex™ LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix
Gibco™

Geltrex™ LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix

Geltrex™ LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-SwarmRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A14133011 mL
A14133025 mL
2 Options
Catalog number A1413301
Price (USD)/ 1 mL
82.25
Add to cart
Quantity:
1 mL
Price (USD)/ 1 mL
82.25
Add to cart
Geltrex™ LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix
Catalog numberA1413301
Price (USD)/ 1 mL
82.25
-
Add to cart
Geltrex™ LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors and is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation. Routinely used for attachment and maintenance of human embryonic stem cells (hESC), every lot of Geltrex™ LDEV-free hESC-qualified Reduced Growth Factor Basement Membrane Matrix is quality control tested using hESCs and StemPro hESC Serum Free Media to help ensure growth and pluripotency of each lot.

Key Features of Geltrex™ LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix:

• Every lot is qualified using hESC, which helps reduce the need for customers to lot screen
• Free of the lactose dehydrogenase elevating virus (LDEV) making it safe for all types of cell culture
• Consistent protein concentration lot-to-lot helps ensure reproducible results

Gain Reliability and Consistency in Your hESC Studies
Human ESCs have specific needs for attachment and maintenance and some lots of basement membrane extract may not provide adequate support. Geltrex™ hESC-qualified reduced growth factor basement membrane matrix has been tested for support of hESC growth and pluripotency, therefore minimizing the need for you to test multiple lots to find one that works. Our consistent manufacturing process helps ensure a protein concentration averaging 15 mg/ml with very little variation from lot-to-lot, so you can have confidence in every purchase.

Free of LDEV Contamination
Recent advances in processing EHS tumor cells have resulted in virus free basement membrane extracts. Every lot is tested by tested by RT-PCR methods so you can now have confidence that every lot of Geltrex™ is virus-free.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHuman induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells
ClassificationReduced Growth Factor
Concentration12 to 18 mg/mL
Culture TypeAdherent Cell Culture
FormFrozen
Product TypeBasement Membrane Matrix
Serum LevelSerum-free
Shelf Life36 months
SterilitySterile
Without AdditivesNo Phenol Red
Endotoxin LevelLow
FormatTube(s)
Product LineGibco™
Purity or Quality GradeCell Culture Grade
Quantity1 mL
Shipping ConditionDry Ice
SpeciesMouse
Unit Size1 mL
Contents & Storage
Geltrex™ LDEV-Free hESC-qualified comes as a sterile protein solution in 1 mL vials.

Store at -20° to -80°C.

Geltrex™ Matrix is stable for 18 months from date of manufacture when properly stored.