Thermo Scientific Chemicals

2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14136.22
100 g, Each
Thermo Scientific Chemicals

2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14136.22
100 g, Each
Quantity
Catalog number: A14136.22
also known as A14136-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41052-75-9
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Assay (Freebase GC)
≥96.0%

Description

2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N'-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N′-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text