Instrument Control Terbium TR-FRET Kit
Instrument Control Terbium TR-FRET Kit
Instrument Control Terbium TR-FRET Kit
The Instrument Control Terbium TR-FRET Kit allows for the rapid assessment of instrument setup for terbium (Tb)-based TR-FRET assays. LanthaScreen™Read more
Have Questions?
Catalog NumberQuantity
A141381 mL kit
Catalog number A14138
Price (USD)
236.00
Each
-
Add to cart
Quantity:
1 mL kit
Price (USD)
236.00
Each
Add to cart
Ask our AI about this Product
The Instrument Control Terbium TR-FRET Kit allows for the rapid assessment of instrument setup for terbium (Tb)-based TR-FRET assays. LanthaScreen™ and Premo™ assays require specific instrument settings. Proper instrument setup is critical to experimental success. This kit contains one tube each of 1x high and low controls which may also be used as positive control samples when running Tb TR-FRET assays.
The Instrument Control Terbium TR-FRET Kit Allows You to:
• Set up instruments for Tb TR-FRET assays quickly
• Check instrument Tb TR-FRET settings
• Read a high and low Tb TR-FRET signal control on each assay plate
Determine if Your Instrument is Set Up to Read Tb TR-FRET Assays
Setting up your instrument to read Tb TR-FRET is the first step in running a successful Tb TR-FRET assay. This kit allows you to quickly determine if your instrument is set up properly to read Tb TR-FRET assays. Simply add a small volume into separate wells, place the plate into the instrument and read. If the ratio of high to low control signal is below 2 the instrument may not be setup properly. If the ratio of high to low control signal is between 2 and 4 you can proceed with a Tb TR-FRET.
Use the Kit as a Positive Control in Tb TR-FRET Assays
When running Tb TR-FRET assays, add a small volume of the high and low controls to separate wells, on your assay plate. Place the plate into the instrument and read, according to instructions. A high to low control signal ratio of between 2 and 4 offers you the confidence that the instrument is performing well.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
For Use With (Equipment)HTS Reader, Microplate Reader
Product LineLanthaScreen™
Quantity1 mL kit
Recommended StorageStore the kit at 4°C
Protect from light
Shipping ConditionWet Ice
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
I have no assay window in my TR-FRET assay. What is the reason?
The single most common reason that a TR-FRET assay fails is that an incorrect choice of emission filters was made. Unlike other fluorescent assays, the filters used in a TR-FRET assay must be exactly those that are recommended for your instrument as the emission filter choice can make or break the assay. The excitation filter has more of an impact on the assay window. Please refer to our instrument setup guides in our instrument compatibility portal (https://www.thermofisher.com/instrumentsetup). If your instrument is not listed there, please contact Drug Discovery Technical Support at drugdiscoverytech@thermofisher.com.
Note: Test your microplate reader's TR-FRET setup before you begin any work with your assay. Using reagents that you have already purchased for your assay, you can test the microplate reader's TR-FRET setup. Please refer to the Terbium (Tb) Assay (http://tools.thermofisher.com/content/sfs/manuals/LanthaScreen_Tb_Instrument_Control_Application_Note17Sep10.pdf) and Europium (Eu) Assay (http://tools.thermofisher.com/content/sfs/manuals/Eu_Instrument_Control_AppNote_12Nov10.pdf) Application Notes for plate reader setup.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
2878583Certificate of AnalysisJan 06, 2024A14138
2756298Certificate of AnalysisJul 13, 2023A14138
2672209Certificate of AnalysisApr 05, 2023A14138
2540535Certificate of AnalysisJul 29, 2022A14138
2507332Certificate of AnalysisJun 24, 2022A14138
5 results displayed, search above for a specific certificate
Safety Data Sheets
Other products to consider
Share catalog number, name or link
1x1 image pixel for data collection