Thermo Scientific Chemicals

Cyclopentanecarbonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14142.06
5 g, Each
Thermo Scientific Chemicals

Cyclopentanecarbonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14142.06
5 g, Each
Quantity
Catalog number: A14142.06
also known as A14142-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4524-93-0
IUPAC Name
cyclopentanecarbonyl chloride
Molecular Formula
C6H9ClO
InChI Key
WEPUZBYKXNKSDH-UHFFFAOYSA-N
SMILES
ClC(=O)C1CCCC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Refractive Index
1.4615-1.4655 @ 20?C

Description

Cyclopentanecarbonyl chloride is used in biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanecarbonyl chloride is used in biochemical for proteomics research.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text