Thermo Scientific™

2-Decanone, 97%, Thermo Scientific Chemicals

Catalog number: A14145.06
5 g, Each
Thermo Scientific™

2-Decanone, 97%, Thermo Scientific Chemicals

Catalog number: A14145.06
5 g, Each
Quantity
Catalog number: A14145.06
also known as A14145-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
693-54-9
IUPAC Name
decan-2-one
Molecular Formula
C10H20O
InChI Key
ZAJNGDIORYACQU-UHFFFAOYSA-N
SMILES
CCCCCCCCC(C)=O
Assay (GC)
>96.0%
Refractive Index
1.4230-1.4280 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text