Thermo Scientific Chemicals

2-Methylpyrazine, 99+%, Thermo Scientific Chemicals

Catalog number: A14146.22
100 g, Each
Thermo Scientific Chemicals

2-Methylpyrazine, 99+%, Thermo Scientific Chemicals

Catalog number: A14146.22
100 g, Each
Quantity
Catalog number: A14146.22
also known as A14146-22
Price (TWD)

Chemical Identifiers

CAS
109-08-0
IUPAC Name
2-methylpyrazine
Molecular Formula
C5H6N2
InChI Key
CAWHJQAVHZEVTJ-UHFFFAOYSA-N
SMILES
CC1=CN=CC=N1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥99.0%
Identification (FTIR)
Conforms
Refractive Index
1.5040-1.5060 @ 20?C

Description

2-Methylpyrazine is used as flavoring agents in bread fresh bread, butter, chocolate, chocolate cocoa, coffee, meat roasted, nut almond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylpyrazine is used as flavoring agents in bread fresh bread, butter, chocolate, chocolate cocoa, coffee, meat roasted, nut almond.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text