Thermo Scientific Chemicals

4-Chloro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals

Catalog number: A14165.14
25 g, Each
Thermo Scientific Chemicals

4-Chloro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals

Catalog number: A14165.14
25 g, Each
Quantity
Catalog number: A14165.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10199-89-0
IUPAC Name
4-chloro-7-nitro-2,1,3-benzoxadiazole
Molecular Formula
C6H2ClN3O3
InChI Key
IGHBXJSNZCFXNK-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(Cl)C2=NON=C12
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Yellow to pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
96-101?C

Description

4-Chloro-7-nitrobenzofurazan is used as a derivatizing reagent for chromatography analysis of amino acids and low molecular weight amines. It is utilized in the preparation of fluorescent phospholipid-derivative, hydroxynaphthofurazan and 4-chloro-7-nitrobenzofurazan- didecanoylphosphatidylethanolamine. It serves as a fluorescent reagent to label free sulfhydryls and N-terminals within proteins and trapping agent for cysteine sulfenic acid. It is a sensitive chromogenic and fluorogenic reagent. It reacts with trans-1-methoxy-3-(trimethylsilyloxy)-1,3-butadiene (Danishefsky's diene) to prepare regioselectively the silyl enol ether. Further, it is used to label peptides, proteins and other biomolecules. In addition it is used in the synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, functionalized hydroxynaphthofurazan and 7-nitrobenzofurazan (NBD)-labeled maleimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-7-nitrobenzofurazan is used as a derivatizing reagent for chromatography analysis of amino acids and low molecular weight amines. It is utilized in the preparation of fluorescent phospholipid-derivative, hydroxynaphthofurazan and 4-chloro-7-nitrobenzofurazan- didecanoylphosphatidylethanolamine. It serves as a fluorescent reagent to label free sulfhydryls and N-terminals within proteins and trapping agent for cysteine sulfenic acid. It is a sensitive chromogenic and fluorogenic reagent. It reacts with trans-1-methoxy-3-(trimethylsilyloxy)-1,3-butadiene (Danishefsky′s diene) to prepare regioselectively the silyl enol ether. Further, it is used to label peptides, proteins and other biomolecules. In addition it is used in the synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, functionalized hydroxynaphthofurazan and 7-nitrobenzofurazan (NBD)-labeled maleimide.

Solubility
Soluble in methanol, dimethylsulfoxide, dimethylformamide and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text