Thermo Scientific Chemicals

6-Aza-2-thiothymine, 98%, Thermo Scientific Chemicals

Catalog number: A14167.06
5 g, Each
Thermo Scientific Chemicals

6-Aza-2-thiothymine, 98%, Thermo Scientific Chemicals

Catalog number: A14167.06
5 g, Each
Quantity
Catalog number: A14167.06
also known as A14167-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-76-9
IUPAC Name
6-methyl-3-sulfanylidene-2,3,4,5-tetrahydro-1,2,4-triazin-5-one
Molecular Formula
C4H5N3OS
InChI Key
NKOPQOSBROLOFP-UHFFFAOYSA-N
SMILES
CC1=NNC(=S)NC1=O
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥97.5 to ≤102.5% (titration)

Description

6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

Solubility
Soluble in water (partly), and formic acid (5 %).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text