6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.
Solubility
Soluble in water (partly), and formic acid (5 %).
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
D I Papac; A Wong; A J Jones. Analysis of acidic oligosaccharides and glycopeptides by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry. Analytical chemistry.1996, 68 (18), 3215-3223.