4-n-Hexylbenzoic acid, 99%, Thermo Scientific Chemicals
4-n-Hexylbenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-n-Hexylbenzoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A14168.03
also known as A14168-03
Price (USD)/ Each
61.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
61.20
Add to cart
4-n-Hexylbenzoic acid, 99%, Thermo Scientific Chemicals
Catalog numberA14168.03
Price (USD)/ Each
61.20
-
Add to cart
Chemical Identifiers
CAS21643-38-9
IUPAC Name4-hexylbenzoic acid
Molecular FormulaC13H18O2
InChI KeyCPEPWESLFZVUEP-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or flakes
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5% to ≤101.5%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5% to ≤101.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only