Thermo Scientific Chemicals

1-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals

Catalog number: A14169.30
250 g, Each
Thermo Scientific Chemicals

1-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals

Catalog number: A14169.30
250 g, Each
Quantity
Catalog number: A14169.30
also known as A14169-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5036-48-6
IUPAC Name
3-(1H-imidazol-1-yl)propan-1-amine
Molecular Formula
C6H11N3
InChI Key
KDHWOCLBMVSZPG-UHFFFAOYSA-N
SMILES
NCCCN1C=CN=C1
Assay (GC)
≥97.5%
Refractive Index
1.5170-1.5210 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid

Description

1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents and air. Air sensitive. Store under dry inert gas. Incompatible with air and carbondioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text