2-Hydroxymethyl-1,4-benzodioxane, 98%, Thermo Scientific Chemicals
2-Hydroxymethyl-1,4-benzodioxane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxymethyl-1,4-benzodioxane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14170.06
also known as A14170-06
Price (USD)/ Each
28.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
28.90
Add to cart
2-Hydroxymethyl-1,4-benzodioxane, 98%, Thermo Scientific Chemicals
Catalog numberA14170.06
Price (USD)/ Each
28.90
-
Add to cart
Chemical Identifiers
CAS3663-82-9
IUPAC Name(2,3-dihydro-1,4-benzodioxin-2-yl)methanol
Molecular FormulaC9H10O3
InChI KeyGWQOQQVKVOOHTI-UHFFFAOYNA-N
SMILESOCC1COC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow to pale orange
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)85.0-92.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only