This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications L-Serine ethyl ester hydrochloride is widely used in pharmaceutical and food industry.
Solubility Soluble in water.
Notes Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.