L-Serine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
L-Serine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Serine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A14174.14
also known as A14174-14
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
Add to cart
L-Serine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Catalog numberA14174.14
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS26348-61-8
IUPAC Namehydrogen ethyl 2-amino-3-hydroxypropanoate chloride
Molecular FormulaC5H12ClNO3
InChI KeyJZJQCLZQSHLSFB-UHFFFAOYNA-N
SMILES[H+].[Cl-].CCOC(=O)C(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)>98.5 to <101.5%
Appearance (Color)White
Optical Rotation-5.0? ±1? (c=10 in Ethanol)
FormCrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Serine ethyl ester hydrochloride is widely used in pharmaceutical and food industry.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
RUO – Research Use Only