Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine sulfate, 98%, Thermo Scientific Chemicals

Catalog number: A14175.22
100 g, Each
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine sulfate, 98%, Thermo Scientific Chemicals

Catalog number: A14175.22
100 g, Each
Quantity
Catalog number: A14175.22
also known as A14175-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
536-47-0
IUPAC Name
N1,N1-dimethylbenzene-1,4-diamine; sulfuric acid
Molecular Formula
C8H14N2O4S
InChI Key
GLUKPDKNLKRLHX-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1
Appearance (Color)
White to brown or grey
Identification (FTIR)
Conforms
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (dry wt. basis)
Loss on Drying
<5.0% (105?/constant wt.)
Form
Powder

Description

N,N-Dimethyl-p-phenylenediamine sulfate salt is used to investigate the role of H2S in cardioprotection induced by ischemic preconditioning in rat heart, in spectrophotometric determination of gold(III) in gold-pharmaceuticals such as sodium aurothimaleate and auranofin, to investigate colonic tissue H2S production using the acetate trapping assay system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-phenylenediamine sulfate salt is used to investigate the role of H2S in cardioprotection induced by ischemic preconditioning in rat heart, in spectrophotometric determination of gold(III) in gold-pharmaceuticals such as sodium aurothimaleate and auranofin, to investigate colonic tissue H2S production using the acetate trapping assay system.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Store in the dark. Incompatible with oxidizing agents and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text