Thermo Scientific Chemicals

4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific Chemicals

Catalog number: A14179.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific Chemicals

Catalog number: A14179.06
5 g, Each
Quantity
Catalog number: A14179.06
also known as A14179-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10558-25-5
IUPAC Name
4-bromo-3,5-dimethyl-1,2-oxazole
Molecular Formula
C5H6BrNO
InChI Key
GYHZPSUAMYIFQD-UHFFFAOYSA-N
SMILES
CC1=C(Br)C(C)=NO1
Appearance (Color)
Clear colorless to yellow to brown
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.4865-1.4915 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text