4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific Chemicals
4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A14179.14
also known as A14179-14
Price (USD)/ Each
654.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
654.00
Add to cart
4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific Chemicals
Catalog numberA14179.14
Price (USD)/ Each
654.00
-
Add to cart
Chemical Identifiers
CAS10558-25-5
IUPAC Name4-bromo-3,5-dimethyl-1,2-oxazole
Molecular FormulaC5H6BrNO
InChI KeyGYHZPSUAMYIFQD-UHFFFAOYSA-N
SMILESCC1=C(Br)C(C)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4865-1.4915 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only