Thermo Scientific Chemicals

Erythrosin B, Thermo Scientific Chemicals

Catalog number: A14180.14
25 g, Each
Thermo Scientific Chemicals

Erythrosin B, Thermo Scientific Chemicals

Catalog number: A14180.14
25 g, Each
Quantity
Catalog number: A14180.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16423-68-0
IUPAC Name
disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Formula
C20H6I4Na2O5
InChI Key
IINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
Appearance (Color)
Red to red-brown
lambda-max (UV-Vis)
524-528nm (water)
Assay (unspecified)
Dye content >= 87.0%
Solution Test
1% in water: clear
Form
Powder

Description

Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

Solubility
Soluble in water, glycerin and methanol.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text