2-Fluorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
2-Fluorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A14181.14
also known as A14181-14
Price (USD)/ Each
424.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
424.00
Add to cart
2-Fluorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA14181.14
Price (USD)/ Each
424.00
-
Add to cart
Chemical Identifiers
CAS2905-21-7
IUPAC Name2-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClFO2S
InChI KeyZSZKAQCISWFDCQ-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid as melt
Refractive Index1.5350-1.5390 @ 20?C
Appearance (Color)Clear, colorless to yellow
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only