Thermo Scientific Chemicals

N-Phenylglycine, 97%

Catalog number: A14182.09
10 g, Each
Thermo Scientific Chemicals

N-Phenylglycine, 97%

Catalog number: A14182.09
10 g, Each
Quantity
Catalog number: A14182.09
also known as A14182-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-01-5
IUPAC Name
sodium 2-(phenylamino)acetate
Molecular Formula
C8H8NNaO2
InChI Key
YEMGQZDWLLBIEY-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)CNC1=CC=CC=C1
Assay (HPLC)
>96.0%
Form
powder
Water Content (Karl Fischer Titration)
<1.0%
Assay (Aqueous acid-base Titration)
>96.0%
Appearance (Color)
Cream to brown

Description

N-Phenylglycine coupled with Glycidyl methacrylate to create a surface active comonomer that promotes adhesive bonding of materials to hard tooth tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylglycine coupled with Glycidyl methacrylate to create a surface active comonomer that promotes adhesive bonding of materials to hard tooth tissues.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text