Thermo Scientific Chemicals

1,3-Dihydroxyacetone dimer, 97%, Thermo Scientific Chemicals

Catalog number: A14189.0B
1000 g, Each
Thermo Scientific Chemicals

1,3-Dihydroxyacetone dimer, 97%, Thermo Scientific Chemicals

Catalog number: A14189.0B
1000 g, Each
Quantity
Catalog number: A14189.0B
also known as A14189-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62147-49-3
Melting Point (clear melt)
75-86?C
Form
Crystals or powder or crystalline powder
Proton NMR
Conforms to structure
Appearance (Color)
White to cream
Assay from Supplier's CofA
≥96.0%

Description

1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text