Thermo Scientific Chemicals

D-Glutamic acid, 99+%

Catalog number: A14191.06
5 g, Each
Thermo Scientific Chemicals

D-Glutamic acid, 99+%

Catalog number: A14191.06
5 g, Each
Quantity
Catalog number: A14191.06
also known as A14191-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
6893-26-1
IUPAC Name
2-aminopentanedioic acid
Molecular Formula
C5H9NO4
InChI Key
WHUUTDBJXJRKMK-UHFFFAOYNA-N
SMILES
NC(CCC(O)=O)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Optical Rotation
-30.0 ? 2.0? (c=10 in 2N HCl)
Assay (Aqueous acid-base Titration)
≥99.0 to ≤101.0%

Description

D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.

Solubility
Soluble in water (8.88 mg/mL) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text