D-Glutamic acid, 99+%
D-Glutamic acid, 99+%
Thermo Scientific Chemicals

D-Glutamic acid, 99+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A14191.14
also known as A14191-14
Price (USD)/ Each
131.65
Online exclusive
146.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
131.65
Online exclusive
146.00 
Save 14.35 (10%)
Add to cart
D-Glutamic acid, 99+%
Catalog numberA14191.14
Price (USD)/ Each
131.65
Online exclusive
146.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6893-26-1
IUPAC Name2-aminopentanedioic acid
Molecular FormulaC5H9NO4
InChI KeyWHUUTDBJXJRKMK-UHFFFAOYNA-N
SMILESNC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Optical Rotation-30.0 ? 2.0? (c=10 in 2N HCl)
Assay (Aqueous acid-base Titration)≥99.0 to ≤101.0%
D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.

Solubility
Soluble in water (8.88 mg/mL) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
Matthew Volgraf; Pau Gorostiza; Rika Numano; Richard H Kramer; Ehud Y Isacoff; Dirk Trauner. Allosteric control of an ionotropic glutamate receptor with an optical switch. Nature Chemical Biology.2006, 1 (2), 47-52.