Thermo Scientific Chemicals

1,3-Benzodioxole, 99%, Thermo Scientific Chemicals

Catalog number: A14192.18
50 g, Each
Thermo Scientific Chemicals

1,3-Benzodioxole, 99%, Thermo Scientific Chemicals

Catalog number: A14192.18
50 g, Each
Quantity
Catalog number: A14192.18
also known as A14192-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
274-09-9
IUPAC Name
2H-1,3-benzodioxole
Molecular Formula
C7H6O2
InChI Key
FTNJQNQLEGKTGD-UHFFFAOYSA-N
SMILES
C1OC2=CC=CC=C2O1
Form
Liquid
Refractive Index
1.5370-1.5400 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥98.5%

Description

1,3-Benzodioxole is useful in gemological stimulant detection. It is also used as a precursor for perfumes, photo initiators, agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzodioxole is useful in gemological stimulant detection. It is also used as a precursor for perfumes, photo initiators, agrochemicals and pharmaceuticals.

Solubility
Soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text