Thermo Scientific Chemicals

D-(+)-Biotin, 98+%, Thermo Scientific Chemicals

Catalog number: A14207.09
10 g, Each
Thermo Scientific Chemicals

D-(+)-Biotin, 98+%, Thermo Scientific Chemicals

Catalog number: A14207.09
10 g, Each
Quantity
Catalog number: A14207.09
also known as A14207-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-85-5
IUPAC Name
5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Molecular Formula
C10H16N2O3S
InChI Key
YBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILES
OC(=O)CCCCC1SCC2NC(=O)NC12
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0% (non-U.S. specification)
Assay from Suppliers CofA
≥98.0% (HPLC, U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Biotin is a form of vitamin B that helps the body to break down fats and carbohydrates. It is used as an alternative medicine to treat or prevent biotin deficiency, which is caused by malnutrition, rapid weight loss, long-term tube feeding and other medical conditions. It is an essential vitamin which is important for amino acid and energy metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot water, dimethyl sulfoxide, alcohol and benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text