p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.
Solubility
Soluble in water, methanol and dimethyl sulfoxide.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- B Toth; and D Nagel. Studies of the tumorigenic potential of 4-substituted phenylhydrazines by the subcutaneous route. Journal of Toxicology and Environmental Health, Part A. 1981, 8 (1-2), 1-9.
- B. Toth; A. Tompa; K. Patill. Tumorigenic effect of 4-methylphenylhydrazine hydrochloride in Swiss mice. Zeitschrift für Krebsforschung und Klinische Onkologie. 1977, 89 (3), 245-252.