Thermo Scientific Chemicals

p-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A14220.14
25 g, Each
Thermo Scientific Chemicals

p-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A14220.14
25 g, Each
Quantity
Catalog number: A14220.14
also known as A14220-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
637-60-5
IUPAC Name
hydrogen (4-methylphenyl)hydrazine chloride
Molecular Formula
C7H11ClN2
InChI Key
HMHWNJGOHUYVMD-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=CC=C(NN)C=C1
Form
Powder
Assay (Titration ex Chloride)
>97.5 to <102.5%
Assay (Freebase GC)
>97.5%
Appearance (Color)
White to pale brown

Description

p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.

Solubility
Soluble in water, methanol and dimethyl sulfoxide.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text