Thermo Scientific Chemicals

Cyclopentanone, 99%, Thermo Scientific Chemicals

Catalog number: A14222.AP
500 mL, Each
Thermo Scientific Chemicals

Cyclopentanone, 99%, Thermo Scientific Chemicals

Catalog number: A14222.AP
500 mL, Each
Quantity
Catalog number: A14222.AP
also known as A14222-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-92-3
IUPAC Name
cyclopentanone
Molecular Formula
C5H8O
InChI Key
BGTOWKSIORTVQH-UHFFFAOYSA-N
SMILES
O=C1CCCC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4355-1.4385 @ 20?C

Description

Cyclopentanone is used as an intermediate in the synthesis of rubber adhesives, synthetic resins, pharmaceuticals and biologically active compounds. It acts as precursor for the preparation of cyclopentamine and also pentethylcyclanone, cyclopentobarbital. It is a useful laboratory reagent and is used as thinner for epoxies. It can also be used as a solvent in paint and varnish removers and for electronic applications. As a dry cleaning agent, it is used for oil extraction. It is also involved in the preparation of cyclopentanone derivatives like cyclopenylamine and cyclopentanol which find application in the perfume industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanone is used as an intermediate in the synthesis of rubber adhesives, synthetic resins, pharmaceuticals and biologically active compounds. It acts as precursor for the preparation of cyclopentamine and also pentethylcyclanone, cyclopentobarbital. It is a useful laboratory reagent and is used as thinner for epoxies. It can also be used as a solvent in paint and varnish removers and for electronic applications. As a dry cleaning agent, it is used for oil extraction. It is also involved in the preparation of cyclopentanone derivatives like cyclopenylamine and cyclopentanol which find application in the perfume industry.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong reducing agents, strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text