Thermo Scientific Chemicals

3-Ethoxy-2-cyclohexen-1-one, 99%, Thermo Scientific Chemicals

Catalog number: A14224.18
50 g, Each
Thermo Scientific Chemicals

3-Ethoxy-2-cyclohexen-1-one, 99%, Thermo Scientific Chemicals

Catalog number: A14224.18
50 g, Each
Quantity
Catalog number: A14224.18
also known as A14224-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5323-87-5
Assay (GC)
≥98.5%
Refractive Index
1.5035-1.5065 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text