Thermo Scientific Chemicals

Sebacoyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A14226.14
25 g, Each
Thermo Scientific Chemicals

Sebacoyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A14226.14
25 g, Each
Quantity
Catalog number: A14226.14
also known as A14226-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-19-3
IUPAC Name
decanedioyl dichloride
Molecular Formula
C10H16Cl2O2
InChI Key
WMPOZLHMGVKUEJ-UHFFFAOYSA-N
SMILES
ClC(=O)CCCCCCCCC(Cl)=O
Appearance (Color)
Clear colorless to white to yellow to pink to brown
Form
Liquid
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%
Identification (FTIR)
Conforms
Refractive Index
1.4670-1.4720 @ 20?C

Description

Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

Solubility
Miscible with hydrocarbons and ethers.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text